Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Cyclobutanetetracarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C25021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25021G Supplier TCI America Supplier No. C25021G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity acid intended for laboratory research applications.

Chemical Identifiers

CAS 53159-92-5
Molecular Formula C8H8O8
Molecular Weight (g/mol) 232.144
MDL Number MFCD00013270
InChI Key CURBACXRQKTCKZ-UHFFFAOYSA-N
PubChem CID 266034
IUPAC Name cyclobutane-1,2,3,4-tetracarboxylic acid
SMILES C1(C(C(C1C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 239°C
Color White-Yellow
Quantity 1 g
Formula Weight 232.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Cyclobutanetetracarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.