Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Cyclopentanetetracarboxylic Dianhydride, MP Biomedicals

Catalog No. ICN220475 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220475 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220475 Supplier MP Biomedicals, Inc Supplier No. 0522047510

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6053-68-5
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
InChI Key NLWBEORDOPDUPM-UHFFFAOYNA-N
Synonym 1,2,3,4-cyclopentanetetracarboxylic dianhydride, dihydro-1h-cyclopenta 1,2-c:3,4-cdifuran-1,3,4,6 3ah,3bh,6ah-tetraone, 1h-cyclopenta 1,2-c:3,4-cdifuran-1,3,4,6 3ah-tetrone, tetrahydro, nlwbeordopdupm-uhfffaoysa-n, 1h-cyclopenta 1,2-c:3,4-c' difuran-1,3,4,6 3ah-tetrone, tetrahydro, ?-2,2,5,5-tetrone, r-1,c-2,c-3,c-4-cyclopentane-1,2:3,4-tetracarboxylic dianhydride, 1h-cyclopenta 1,2-c:3,4-c' difuran-1,3,4,6 3ah-tetrone, tetrahydro-, 3aalpha,3balpha,6aalpha,7aalpha, 1,2,3,4-cyclopentanetetracarboxylic 1,2:3,4-dianhydride, 4,10-dioxatricyclo 6.3.0.0<2,6> undecane-3,5,9,11-tetraone
PubChem CID 110757

Specifications

Quantity 10 g
Formula Weight 210.14
Chemical Name or Material 1,2,3,4-Cyclopentanetetracarboxylic Dianhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.