Learn More
Medchemexpress LLC 1,2:3,4-di-O-isopropylidene-α-D-galactopyranose | 4064-06-6 | MFCD00063225 | 97.0% | 260.28 g/mol | C12H20O6 | 100 G
SDP
Catalog No. 5000299738
Encompass_Preferred
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is a protected D-galactopyranose derivative (diacetonide) used as a carbohydrate building block and biochemical reagent. The acetal-protected sugar acts as a stable intermediate for selective transformations and glycoconjugate assembly in research and synthetic workflows.
  • Protected sugar acetal suitable for selective functionalization.
  • Used as a synthetic intermediate in carbohydrate chemistry.
  • Compatible with common organic reaction and deprotection conditions.
  • Provided with analytical data for quality verification.
  • Available in multiple pack sizes for research applications.
Catalog No. 50-002-99738 Supplier Medchemexpress LLC Supplier No. HYW036647100G
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is a protected D-galactopyranose derivative (diacetonide) used as a carbohydrate building block and biochemical reagent. The acetal-protected sugar acts as a stable intermediate for selective transformations and glycoconjugate assembly in research and synthetic workflows.
  • Protected sugar acetal suitable for selective functionalization.
  • Used as a synthetic intermediate in carbohydrate chemistry.
  • Compatible with common organic reaction and deprotection conditions.
  • Provided with analytical data for quality verification.
  • Available in multiple pack sizes for research applications.