Learn More
Medchemexpress LLC 1,2:3,4-di-O-isopropylidene-α-D-galactopyranose | 4064-06-6 | MFCD00063225 | 97.0% | 260.28 g·mol⁻¹ | C12H20O6 | 25 G
SDP
Catalog No. 5000303024
Encompass_Preferred
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is an organic reagent used to provide a protected form of D-galactopyranose for carbohydrate chemistry and multi-step synthesis. It is supplied in a high-purity form suitable for research applications and as an intermediate in synthetic workflows.
  • Provides isopropylidene protection for the 1,2 and 3,4 hydroxyl groups of D-galactopyranose.
  • Useful intermediate in synthesis of carbohydrates and glycosylation studies.
  • High purity suitable for research-grade synthetic applications.
  • Supplied as a colorless to light yellow liquid with reported density 1.143 g·cm⁻3.
  • Available in multiple pack sizes for laboratory and bulk use.
  • Compatible with standard protecting group and deprotection protocols.
Catalog No. 50-003-03024 Supplier Medchemexpress LLC Supplier No. 25GHYW036647
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is an organic reagent used to provide a protected form of D-galactopyranose for carbohydrate chemistry and multi-step synthesis. It is supplied in a high-purity form suitable for research applications and as an intermediate in synthetic workflows.
  • Provides isopropylidene protection for the 1,2 and 3,4 hydroxyl groups of D-galactopyranose.
  • Useful intermediate in synthesis of carbohydrates and glycosylation studies.
  • High purity suitable for research-grade synthetic applications.
  • Supplied as a colorless to light yellow liquid with reported density 1.143 g·cm⁻3.
  • Available in multiple pack sizes for laboratory and bulk use.
  • Compatible with standard protecting group and deprotection protocols.