Learn More
Medchemexpress LLC 1,2:3,4-di-O-isopropylidene-α-D-galactopyranose | 4064-06-6 | MFCD00063225 | 97.0% | 260.28 g/mol | C12H20O6 | 50 G
SDP
Catalog No. 5000301191
Encompass_Preferred
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is an isopropylidene-protected derivative of D-galactopyranose used as a synthetic building block and reagent in carbohydrate chemistry and glycobiology research. It is employed to protect vicinal diols during multistep syntheses, enabling selective modifications and assembly of oligosaccharides.
  • Protects 1,2- and 3,4-hydroxyl groups for selective carbohydrate transformations.
  • Useful building block for oligosaccharide synthesis and glycoconjugate preparation.
  • Available as a high-purity reagent suitable for analytical and synthetic work.
  • Soluble in common organic solvents used in synthetic protocols.
  • Facilitates regioselective functionalization of galactose derivatives.
Catalog No. 50-003-01191 Supplier Medchemexpress LLC Supplier No. HYW03664750G
1,2:3,4-Di-O-isopropylidene-α-D-galactopyranose is an isopropylidene-protected derivative of D-galactopyranose used as a synthetic building block and reagent in carbohydrate chemistry and glycobiology research. It is employed to protect vicinal diols during multistep syntheses, enabling selective modifications and assembly of oligosaccharides.
  • Protects 1,2- and 3,4-hydroxyl groups for selective carbohydrate transformations.
  • Useful building block for oligosaccharide synthesis and glycoconjugate preparation.
  • Available as a high-purity reagent suitable for analytical and synthetic work.
  • Soluble in common organic solvents used in synthetic protocols.
  • Facilitates regioselective functionalization of galactose derivatives.