Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydro-1-naphthylamine, TCI America™
SDP

Catalog No. T25015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25015G Supplier TCI America Supplier No. T25015G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2217-40-5
Molecular Formula C10H14ClN
Molecular Weight (g/mol) 183.68
MDL Number MFCD00001740
InChI Key DETWFIUAXSWCIK-UHFFFAOYNA-N
Synonym 1,2,3,4-tetrahydro-1-naphthylamine, 1-aminotetralin, aminotetralin, 1-amino-1,2,3,4-tetrahydro-naphthalene, aminotetralin czech, tetrahydro-beta-naphthylamine, 1-amino-1,2,3,4-tetrahydronaphthalene, 1-naphthalenamine, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthylamine, beta-1,2,3,4-tetrahydronaphthylamine
PubChem CID 18066
IUPAC Name 1,2,3,4-tetrahydronaphthalen-1-aminium chloride
SMILES [Cl-].[NH3+]C1CCCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 230°C
Quantity 5 g
Formula Weight 147.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3,4-Tetrahydro-1-naphthylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.