Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydro-2-naphthol, MP Biomedicals

Catalog No. ICN205634 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205634 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205634 Supplier MP Biomedicals, Inc Supplier No. 0520563405

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 529-33-9
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00001739,MFCD00063006
InChI Key JAAJQSRLGAYGKZ-UHFFFAOYNA-N
Synonym 1,2,3,4-tetrahydro-1-naphthol, tetralin-1-ol, 1-tetralol, alpha-tetralol, 1-hydroxytetralin, 1-naphthalenol, 1,2,3,4-tetrahydro, .alpha.-tetralol, tetrahydro-1-naphthol van, +-1,2,3,4-tetrahydro-1-naphthol, tetrahydro-1-naphthol
PubChem CID 10723
IUPAC Name 1,2,3,4-tetrahydronaphthalen-1-ol
SMILES OC1CCCC2=CC=CC=C12

Specifications

Quantity 5 g
Formula Weight 148.21
Chemical Name or Material 1,2,3,4-Tetrahydro-2-Naphthol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.