Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydrobenzazepine-5-one, 97%, Thermo Scientific™

Catalog No. AC435910010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC435910010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435910010 Supplier Thermo Scientific Chemicals Supplier No. 435910010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1127-74-8
MDL Number MFCD03426404
InChI Key NKRKBYFBKLDCFB-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-benzo b azepin-5-one, 1,2,3,4-tetrahydrobenzo b azepin-5-one, 1,2,3,4-tetrahydrobenzoazepine-5-one, 3,4-dihydro-1h-benzo b azepin-5 2h-one, 1,2,3,4-tetrahydro-5h-1-benzazepin-5-one, 2,3,4,5-tetrahydro-1h-1-benzazepin-5-one, 5h-1-benzazepin-5-one, 1,2,3,4-tetrahydro, 5-oxo-2,3,4,5-tetrahydro-1h-benzazepine, 1,2,3,4-tetrahydrobenzazepine-5-one, zlchem 1002
PubChem CID 294788
IUPAC Name 1,2,3,4-tetrahydro-1-benzazepin-5-one
SMILES C1CC(=O)C2=CC=CC=C2NC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 71.0°C
Color Green to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H11NO
Quantity 1 g
Molecular Weight (g/mol) 161.21
Formula Weight 161.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,2,3,4-Tetrahydrobenzazepine-5-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.