Learn More
Medchemexpress LLC 1,2:3,5-di-O-isopropylidene-alpha-D-xylofuranose | 20881-04-3 | MFCD03002332 | 98.9% | 230.26 g/mol | C11H18O5 | 1 G
SDP
Catalog No. 5000283762
Encompass_Preferred
1,2:3,5-Di-O-isopropylidene-α-D-xylofuranose is a protected sugar derivative used as an intermediate and reagent in organic synthesis and carbohydrate chemistry. It is supplied as a white to off-white solid suitable for laboratory research and synthesis applications.
  • Useful building block in carbohydrate and nucleoside synthesis.
  • Acetal protection (isopropylidene) improves stability and chemoselectivity.
  • High purity (98.9%) for reliable reaction performance.
  • White to off-white solid, easy to handle and store.
  • Molecular formula C11H18O5; CAS number 20881-04-3.
  • Available in small research pack sizes, e.g. 1 g.
Catalog No. 50-002-83762 Supplier Medchemexpress LLC Supplier No. HYW1455751G
1,2:3,5-Di-O-isopropylidene-α-D-xylofuranose is a protected sugar derivative used as an intermediate and reagent in organic synthesis and carbohydrate chemistry. It is supplied as a white to off-white solid suitable for laboratory research and synthesis applications.
  • Useful building block in carbohydrate and nucleoside synthesis.
  • Acetal protection (isopropylidene) improves stability and chemoselectivity.
  • High purity (98.9%) for reliable reaction performance.
  • White to off-white solid, easy to handle and store.
  • Molecular formula C11H18O5; CAS number 20881-04-3.
  • Available in small research pack sizes, e.g. 1 g.