Learn More
Medchemexpress LLC 1,2:3,5-di-O-isopropylidene-α-D-xylofuranose | 20881-04-3 | MFCD00063224 | 98.9% | 230.26 g/mol | C11H18O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1455755G
Encompass_Preferred
1,2:3,5-Di-O-isopropylidene-α-D-xylofuranose is an isopropylidene-protected derivative of α-D-xylofuranose supplied as a white to off-white solid. It is used as a protected sugar intermediate and building block in organic synthesis and carbohydrate chemistry, suitable for laboratory research and small-scale synthesis.
  • Protected sugar intermediate for synthetic chemistry.
  • High purity (approx. 98.9%) suitable for research applications.
  • White to off-white solid for convenient handling and storage.
  • Compatible with common carbohydrate modification and derivatization reactions.
  • Available in typical laboratory pack sizes for small-scale use.
Catalog No. 50-002-83718