Learn More
Medchemexpress LLC HY-111832 5mg Medchemexpress, 1,2,3,6-Tetragalloylglucose CAS:79886-50-3 Purity:>98%
SDP
Catalog No. 501968977
Encompass_Preferred
Medchemexpress, HY-111832 5mg 1,2,3,6-Tetragalloylglucose CAS:79886-50-3 1,2,3,6-Tetragalloylglucose is a potent UDP glucuronosyltransferase 1 family, polypeptide A1 (UGT1A1) inhibitor, with a Ki of 1.68 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8977 Supplier Medchemexpress LLC Supplier No. HY1118325MG
Medchemexpress, HY-111832 5mg 1,2,3,6-Tetragalloylglucose CAS:79886-50-3 1,2,3,6-Tetragalloylglucose is a potent UDP glucuronosyltransferase 1 family, polypeptide A1 (UGT1A1) inhibitor, with a Ki of 1.68 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.