Learn More
Cayman Chemical 1,2,3,7,8,9-Hexachlorodibenzo-p-dioxin, Purity: ≥90%, CAS: 19408-74-3, 1 mg
SDP
Catalog No. 502919047
Encompass_Preferred
A polychlorinated dibenzodioxin; induces the formation of liver adenomas or carcinomas in rats at 2.5 µg/kg once per week in combination with 1,2,3,6,7,8-hexachlorodibenzo-p-dioxin; has been found in the air near municipal waste incinerators and in blood from individuals that lived near or in herbicide-sprayed regions
Catalog No. 50-291-9047 Supplier Cayman Chemical Supplier No. 385961 MG
A polychlorinated dibenzodioxin; induces the formation of liver adenomas or carcinomas in rats at 2.5 µg/kg once per week in combination with 1,2,3,6,7,8-hexachlorodibenzo-p-dioxin; has been found in the air near municipal waste incinerators and in blood from individuals that lived near or in herbicide-sprayed regions