Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Benzothiadiazole-5-carbonyl chloride, 95%, Thermo Scientific™

Catalog No. CC09402DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09402DA 1 g
CC09402CB 250 mg
CC09402DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC09402DA Supplier Thermo Scientific Chemicals Supplier No. CC09402DA
Add to Cart

Chemical Identifiers

CAS 321309-32-4
Molecular Formula C7H3ClN2OS
Molecular Weight (g/mol) 198.624
InChI Key VOSTUOLSYXVSND-UHFFFAOYSA-N
Synonym 1,2,3-benzothiadiazole-5-carbonylchloride, benzo d 1,2,3-thiadiazole-5-carbonyl chloride, benzo d 1,2,3 thiadiazole-5-carbonyl chloride
PubChem CID 2735457
IUPAC Name 1,2,3-benzothiadiazole-5-carbonyl chloride
SMILES C1=CC2=C(C=C1C(=O)Cl)N=NS2

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 198.63
Percent Purity 95%
Chemical Name or Material 1, 2, 3-Benzothiadiazole-5-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.