Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,3-Dichlorophenyl)ethan-1-one, 97%, Thermo Scientific™

Catalog No. TL00325DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
TL00325DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00325DA Supplier Thermo Scientific Chemicals Supplier No. TL00325DA

Specifications

CAS 56041-57-7
CAS Min % 97.0
Density 1.293g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6Cl2O
Linear Formula Cl2 C6 H3 C (O) C H3
Quantity 1g
MDL Number MFCD00052988
Synonym 1-2,3-dichlorophenyl ethanone, 2',3'-dichloroacetophenone, 2,3-dichloroacetophenone, 1-2,3-dichlorophenyl ethan-1-one, ethanone, 1-2,3-dichlorophenyl, 1-acetyl-2,3-dichlorobenzene, pubchem3368, 1-2,3-dichlorophenyl-1-ethanone, 1-2,3-bis chloranyl phenyl ethanone
InChI Key KMABBMYSEVZARZ-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC(Cl)=C1Cl
IUPAC Name 1-(2,3-dichlorophenyl)ethan-1-one
Molecular Weight (g/mol) 189.04
PubChem CID 91986
Formula Weight 189.04
Percent Purity 97%
Chemical Name or Material 1-(2, 3-Dichlorophenyl)ethan-1-one
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.