Learn More
Medchemexpress LLC 1-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-phenylbutane-1,4-dione | 1360540-81-3 | 98.2% | C18H16O4 | 10MG
SDP
Catalog No. 5000260787
Encompass_Preferred
Dalosirvat is a small-molecule Wnt pathway activator originally described in patent WO2012024404A1. It is supplied for research use and exhibits potent Wnt activation with reported EC50 values of 28-29 nM. The compound is provided as a light-yellow solid, has molecular formula C18H16O4, and is characterized with high purity for laboratory studies.
  • Potent Wnt pathway activation (EC50 28-29 nM).
  • High purity (~98.2%) for reproducible results.
  • Available as solid and as a DMSO solution for flexible use.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for in vitro research applications targeting Wnt signaling.
Catalog No. 50-002-60787 Supplier Medchemexpress LLC Supplier No. HY13551610MG
Dalosirvat is a small-molecule Wnt pathway activator originally described in patent WO2012024404A1. It is supplied for research use and exhibits potent Wnt activation with reported EC50 values of 28-29 nM. The compound is provided as a light-yellow solid, has molecular formula C18H16O4, and is characterized with high purity for laboratory studies.
  • Potent Wnt pathway activation (EC50 28-29 nM).
  • High purity (~98.2%) for reproducible results.
  • Available as solid and as a DMSO solution for flexible use.
  • Stable under recommended storage conditions for long-term use.
  • Suitable for in vitro research applications targeting Wnt signaling.