Learn More
Medchemexpress LLC 1,2,3-propanetricarboxylic acid | 99-14-9 | MFCD00002723 | 176.12 g/mol | C6H8O6 | 1 ML
SDP
Catalog No. 5000300881
Encompass_Preferred
Tricarballylic acid (1,2,3-propanetricarboxylic acid) is a small-molecule research reagent (CAS 99-14-9) used as an aconitate hydratase inhibitor and analytical standard. It is commonly supplied as a ready-to-use solution for biochemical assays and metabolic studies; molecular formula C6H8O6 and molecular weight 176.12 g/mol.
  • Used as an aconitate hydratase inhibitor in biochemical assays.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid use.
  • Chemical identifiers include CAS 99-14-9 and MFCD00002723 for database reference.
  • Molecular formula C6H8O6 and molecular weight 176.12 g/mol support analytical calculations.
  • Reported to chelate magnesium and be applicable in metabolic pathway studies.
Catalog No. 50-003-00881 Supplier Medchemexpress LLC Supplier No. HYW02021510MM/1ML
Tricarballylic acid (1,2,3-propanetricarboxylic acid) is a small-molecule research reagent (CAS 99-14-9) used as an aconitate hydratase inhibitor and analytical standard. It is commonly supplied as a ready-to-use solution for biochemical assays and metabolic studies; molecular formula C6H8O6 and molecular weight 176.12 g/mol.
  • Used as an aconitate hydratase inhibitor in biochemical assays.
  • Available as a ready-to-use 10 mM solution in DMSO for rapid use.
  • Chemical identifiers include CAS 99-14-9 and MFCD00002723 for database reference.
  • Molecular formula C6H8O6 and molecular weight 176.12 g/mol support analytical calculations.
  • Reported to chelate magnesium and be applicable in metabolic pathway studies.