Learn More
Medchemexpress LLC 1,2,3-Propanetriol, 1,2,3-triacetate | 102-76-1 | MFCD00008716 | 99.87% | 250 MG
SDP
Catalog No. 5000405608
Encompass_Preferred
Triacetin (Standard) is the analytical standard for Triacetin. Intended for research and analytical applications, this synthetic compound is a triester of glycerol and acetic acid.
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC applications
  • Suitable for GC applications
  • Suitable for MS applications
Catalog No. 50-004-05608 Supplier Medchemexpress LLC Supplier No. 250MGHYB0896R
Triacetin (Standard) is the analytical standard for Triacetin. Intended for research and analytical applications, this synthetic compound is a triester of glycerol and acetic acid.
  • Reference standard supplied assay
  • Commonly used in qualitative research experiments
  • Commonly used in quantitative research experiments
  • Commonly used in methodological research experiments
  • Suitable for HPLC applications
  • Suitable for GC applications
  • Suitable for MS applications