Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Thiadiazole-4-carbaldehyde, Thermo Scientific™

Catalog No. SP01373DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP01373DE 5 g Amber Glass Bottle
SP01373CB 250 mg Amber Glass Bottle
SP01373DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SP01373DE Supplier Thermo Scientific Chemicals Supplier No. SP01373DE
Add to Cart

Chemical Identifiers

CAS 27643-15-8
Molecular Formula C3H2N2OS
Molecular Weight (g/mol) 114.122
InChI Key JNEBZFFTOLBIKJ-UHFFFAOYSA-N
Synonym 1,2,3-thiadiazole-4-carbaldehyde, 1,2,3-thiadiazole-4-carboxaldehyde, 4-thiadiazolecarboxaldehyde, acmc-1cb1x, 1,2,3-thiadiazol-4-carboxaldehyde
PubChem CID 2737271
IUPAC Name thiadiazole-4-carbaldehyde
SMILES C1=C(N=NS1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 114.13
Percent Purity 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.