Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Tribromopropane 97.0+%, TCI America™
SDP

Catalog No. T035525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T035525G 25 g
T0355500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T035525G Supplier TCI America Supplier No. T035525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96-11-7
Molecular Formula C3H5Br3
Molecular Weight (g/mol) 280.79
MDL Number MFCD00017884
InChI Key FHCLGDLYRUPKAM-UHFFFAOYSA-N
Synonym s-tribromopropane, glyceryl tribromohydrin, sym-tribromopropane, glycerol tribromohydrin, propane, 1,2,3-tribromo, unii-d2r8l96tov, ccris 6706, d2r8l96tov, 1,3-tribromopropane, propane,2,3-tribromo
PubChem CID 7279
ChEBI CHEBI:18859
IUPAC Name 1,2,3-tribromopropane
SMILES BrCC(Br)CBr

Specifications

Melting Point 17°C
Color Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 280.79
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Tribromopropane
EINECSNumber (2)-0084
RTECSNumber TZ8300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.