Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropane, MilliporeSigma™ Supelco™

Catalog No. 111020491 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0491 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0491 Supplier MilliporeSigma Supelco Supplier No. 47794
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 96-18-4
Density 1.387 g/mL (at 25°C)
Flash Point 74°C
Physical Form Neat
Linear Formula CH2 ClCHClCH2 Cl
MDL Number MFCD00000946
Melting Point -14°C
Boiling Point 156°C
Synonym Glycerol trichlorohydrin; Trichlorohydrin
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 147.43
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.