Learn More
Cayman Chemical 1-(2,3-Dichlorophenyl)urea, Purity: ≥95%, CAS: 13142-51-3, 250 mg
SDP
Catalog No. 502916563
Encompass_Preferred
A building block; has been used in the synthesis of compounds with cytokinin-like activity
Catalog No. 50-291-6563 Supplier Cayman Chemical Supplier No. 39160250 MG
A building block; has been used in the synthesis of compounds with cytokinin-like activity