Learn More
Cayman Chemical 1 2 3TrI10 ZHeptadecenoyl 10mg
SDP
Catalog No. 502938422
Encompass_Preferred
An internal standard for the quantification of 1,2,3-tri-10(Z)-heptadecenoyl glycerol by GC- or LC-MS
Catalog No. 50-293-8422 Supplier Cayman Chemical Supplier No. 2892010 MG
An internal standard for the quantification of 1,2,3-tri-10(Z)-heptadecenoyl glycerol by GC- or LC-MS