Learn More
Cayman Chemical 1,2,3-Tri-13(E)-Docosenoyl Glycerol, Purity: ≥98%, CAS: 37635-44-2, 100 mg
SDP
Catalog No. 502941624
Encompass_Preferred
A triacylglycerol; dietary administration at 15% of a calcium-deficient diet transiently increases heart triglyceride levels, which return to baseline after 28 days
Catalog No. 50-294-1624 Supplier Cayman Chemical Supplier No. 26991100 MG
A triacylglycerol; dietary administration at 15% of a calcium-deficient diet transiently increases heart triglyceride levels, which return to baseline after 28 days