Learn More
Cayman Chemical 1,2,3-Tri-13(Z)-Docosenoyl Glycerol, Purity: ≥95%, CAS: 2752-99-0, 250 mg
SDP
Catalog No. 502934621
Encompass_Preferred
A triacylglycerol; has been found in nasturtium seed oil; dietary administration at 30% of fat calories in a 60% fat diet, in combination with sunflowerseed oil, induces severe cardiac lipidosis in rats after 1 week, with severity decreasing by 24 weeks; is a minor component of Lorenzo’s oil
Catalog No. 50-293-4621 Supplier Cayman Chemical Supplier No. 26894250 MG
A triacylglycerol; has been found in nasturtium seed oil; dietary administration at 30% of fat calories in a 60% fat diet, in combination with sunflowerseed oil, induces severe cardiac lipidosis in rats after 1 week, with severity decreasing by 24 weeks; is a minor component of Lorenzo’s oil