Learn More
Cayman Chemical 1,2,3-Tri-α-Linolenoyl Glycerol, Purity: ≥98%, CAS: 14465-68-0, 50 mg
SDP
Catalog No. 502891418
Encompass_Preferred
A polyunsaturated triacylglycerol with α-linolenoyl (18:3) side-chains attached at the C-1, C-2, and C-3 positions that is useful in lipid research
Catalog No. 50-289-1418 Supplier Cayman Chemical Supplier No. 1000975850 MG
A polyunsaturated triacylglycerol with α-linolenoyl (18:3) side-chains attached at the C-1, C-2, and C-3 positions that is useful in lipid research