Learn More
Cayman Chemical 1,2,3-Triarachidonoyl Glycerol, Purity: ≥98%, CAS: 23314-57-0, 10 mg
SDP
Catalog No. 502890132
Encompass_Preferred
A polyunsaturated triacylglycerol with arachidonoyl (20:4) side-chains attached at the C-1, C-2, and C-3 positions that is useful in lipid research
Catalog No. 50-289-0132 Supplier Cayman Chemical Supplier No. 1000982610 MG
A polyunsaturated triacylglycerol with arachidonoyl (20:4) side-chains attached at the C-1, C-2, and C-3 positions that is useful in lipid research