Learn More
Cayman Chemical 1 2 3TrIdocosahexaenoyl Glyc
SDP
Catalog No. 502993795
Encompass_Preferred
A selective inhibitor of mutant EGFR that exhibits a 298-fold increase in potency compared with wild-type EGFR oral administration at 500 mg/kg regresses tumors with EGFR-active and T790M mutations in a xenograft model
Catalog No. 50-299-3795 Supplier Cayman Chemical Supplier No. 23152100 MG
A selective inhibitor of mutant EGFR that exhibits a 298-fold increase in potency compared with wild-type EGFR oral administration at 500 mg/kg regresses tumors with EGFR-active and T790M mutations in a xenograft model