Learn More
Cayman Chemical 1 2 3TrIdocosahexaenoyl Glyc
SDP
Supplier:  Cayman Chemical 23152250 MG
Encompass_Preferred
A selective inhibitor of mutant EGFR that exhibits a 298-fold increase in potency compared with wild-type EGFR oral administration at 500 mg/kg regresses tumors with EGFR-active and T790M mutations in a xenograft model
Catalog No. 50-298-7599
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.