Learn More
Cayman Chemical 1,2,3-Trielaidoyl Glycerol, Purity: ≥95%, CAS: 537-39-3, 100 mg
SDP
Catalog No. 502941398
Encompass_Preferred
A triacylglycerol; dietary administration reduces serum cholesterol levels in rats compared to corn oil-fed control animals; has been used as a substrate to determine the substrate specificity of S. rimosus extracellular lipase
Catalog No. 50-294-1398 Supplier Cayman Chemical Supplier No. 26950100 MG
A triacylglycerol; dietary administration reduces serum cholesterol levels in rats compared to corn oil-fed control animals; has been used as a substrate to determine the substrate specificity of S. rimosus extracellular lipase