Learn More
Cayman Chemical 1,2,3-Triheneicosanoyl Glycerol, Purity: ≥95%, CAS: 26536-14-1, 50 mg
SDP
Catalog No. 502890161
Encompass_Preferred
A triacylglycerol containing heneicosanoic acid groups at the sn-1, sn-2, and sn-3 positions; has been used as an internal standard for the quantification of fatty acids in human breast milk samples and triacylglycerols in rat heart
Catalog No. 50-289-0161 Supplier Cayman Chemical Supplier No. 1001054050 MG
A triacylglycerol containing heneicosanoic acid groups at the sn-1, sn-2, and sn-3 positions; has been used as an internal standard for the quantification of fatty acids in human breast milk samples and triacylglycerols in rat heart