Learn More
Cayman Chemical 1,2,3-Trilinoelaidoyl Glycerol, Purity: ≥98%, CAS: 5188-25-0, 100 mg
SDP
Catalog No. 502941637
Encompass_Preferred
A triacylglycerol; dietary administration lowers serum levels of TXB2, PGF2, and PGE in rats from 0.6-6.3% w/w
Catalog No. 50-294-1637 Supplier Cayman Chemical Supplier No. 26948100 MG
A triacylglycerol; dietary administration lowers serum levels of TXB2, PGF2, and PGE in rats from 0.6-6.3% w/w