Learn More
Cayman Chemical 1,2,3-Trinonadecanoyl Glycerol, Purity: ≥98%, CAS: 26536-13-0, 1 g
SDP
Catalog No. 502937640
Encompass_Preferred
A triacylglycerol; has been used as an internal standard for the quantification of triacylglycerols in human serum by capillary gas chromatography
Catalog No. 50-293-7640 Supplier Cayman Chemical Supplier No. 268981 G
A triacylglycerol; has been used as an internal standard for the quantification of triacylglycerols in human serum by capillary gas chromatography