Learn More
Cayman Chemical 1,2,3-Trioctanoyl Glycerol-13C3, Purity: ≥95%, CAS: 65402-55-3, 50 mg
SDP
Catalog No. 502989164
Encompass_Preferred
An internal standard for the quantification of 1,2,3-trioctanoyl glycerol-13C3
Catalog No. 50-298-9164 Supplier Cayman Chemical Supplier No. 2946450 MG
An internal standard for the quantification of 1,2,3-trioctanoyl glycerol-13C3