Learn More
Cayman Chemical 1,2,3-Trioleoyl Glycerol-d5, Purity: ≥99% deuterated forms (d1-d5), CAS: 60892-83-3, 10 mg
SDP
Catalog No. 502943918
Encompass_Preferred
An internal standard for the quantification of 1,2,3-trioleoyl glycerol by GC- or LC-MS
Catalog No. 50-294-3918 Supplier Cayman Chemical Supplier No. 4198710 MG
An internal standard for the quantification of 1,2,3-trioleoyl glycerol by GC- or LC-MS