Learn More
Cayman Chemical 1,2,3-Tripalmitoyl-d31 Glycerol, Purity: ≥99% deuterated forms (d1-d93), CAS: 241157-04-0, 50 mg
SDP
Catalog No. 502938810
Encompass_Preferred
An internal standard for the quantification of 1,2,3-tripalmitoyl glycerol by GC- or LC-MS
Catalog No. 50-293-8810 Supplier Cayman Chemical Supplier No. 2859350 MG
An internal standard for the quantification of 1,2,3-tripalmitoyl glycerol by GC- or LC-MS