Learn More
Cayman Chemical 1,2,3-Tripentadecanoyl Glycerol, Purity: ≥98%, CAS: 7370-46-9, 100 mg
SDP
Catalog No. 502941263
Encompass_Preferred
A triacylglycerol; has been found in various grapes; has been used as a standard for the detection of triacylglycerols in human plasma by LC-MS
Catalog No. 50-294-1263 Supplier Cayman Chemical Supplier No. 26962100 MG
A triacylglycerol; has been found in various grapes; has been used as a standard for the detection of triacylglycerols in human plasma by LC-MS