Learn More
Cayman Chemical 1 2 3TrIstarol GlcRold1 10mg
SDP
Supplier:  Cayman Chemical 2886710 MG
Encompass_Preferred
An internal standard for the quantification of 1,2,3-tristearoyl glycerol by GC- or LC-MS
Catalog No. 50-294-0811