Learn More
Cayman Chemical 1,2,3-Tritridecanoyl Glycerol, Purity: ≥98%, CAS: 26536-12-9, 500 mg
SDP
Catalog No. 502941596
Encompass_Preferred
A synthetic triacylglycerol; has been used as a standard for the relative quantification of triacylglycerols in C. elegans; has been used as an internal standard for the quantification of triacylglycerols in the serum and liver of adult rat offspring receiving dietary conjugated linoleic acids during and after gestation
Catalog No. 50-294-1596 Supplier Cayman Chemical Supplier No. 26899500 MG
A synthetic triacylglycerol; has been used as a standard for the relative quantification of triacylglycerols in C. elegans; has been used as an internal standard for the quantification of triacylglycerols in the serum and liver of adult rat offspring receiving dietary conjugated linoleic acids during and after gestation