Learn More
Cayman Chemical 1 2 3TrIundecnoyl GlycRol 10mg
SDP
Catalog No. 502941733
Encompass_Preferred
A triacylglycerol; dietary administration prevents fasting-induced decreases in plasma glucose, alanine, and insulin levels, as well as the ratio of insulin to glucagon, in rats
Catalog No. 50-294-1733 Supplier Cayman Chemical Supplier No. 2700810 MG
A triacylglycerol; dietary administration prevents fasting-induced decreases in plasma glucose, alanine, and insulin levels, as well as the ratio of insulin to glucagon, in rats