Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Tetrachlorobenzene, 98%

Catalog No. AAA1431830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431830 250 g
AAA1431809 10 g
AAA1431818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AAA1431830 Supplier Thermo Scientific Chemicals Supplier No. A1431830
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 95-94-3 | C6H2Cl4 | 215.882 g/mol

It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a high melting diluent in the eutectic crystallization of pseudo binary systems of polyethylene and high melting diluents.

Solubility
Insoluble in water. Solubility in toluene is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-94-3
Molecular Formula C6H2Cl4
Molecular Weight (g/mol) 215.882
MDL Number MFCD00000549
InChI Key JHBKHLUZVFWLAG-UHFFFAOYSA-N
Synonym s-tetrachlorobenzene, benzene, 1,2,4,5-tetrachloro, benzene tetrachloride, rcra waste number u207, 1,2,4,5-tetrachlorbenzol, unii-5n27529kgh, ccris 766, benzene, tetrachloro, rcra waste no. u207, 1,2,4,5-tetrachlorbenzol russian
PubChem CID 7270
ChEBI CHEBI:36697
IUPAC Name 1,2,4,5-tetrachlorobenzene
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)Cl

Specifications

Melting Point 138°C to 142°C
Density 1.86
Quantity 250 g
Odor Unpleasant
UN Number UN3077
Beilstein 1618315
Boiling Point 245°C
Solubility Information Insoluble in water. Solubility in toluene is almost transparent.
Formula Weight 215.89
Percent Purity 98%
Chemical Name or Material 1,2,4,5-Tetrachlorobenzene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-466-2
RTECSNumber DB9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.