Learn More
Medchemexpress LLC 1,2,4,5-Tetrazine, 3-methyl-6-[4-(3,6,9,12-tetraoxapentadec-14-yn-1-yloxy)phenyl]- | 1802907-97-6 | 98.0% | C20H26N4O5 | 25 MG
SDP
Catalog No. 5000357378
Encompass_Preferred
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest. They function by exploiting the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system
Catalog No. 50-003-57378 Supplier Medchemexpress LLC Supplier No. 25MGHY141275
Methyltetrazine-PEG5-alkyne is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are molecules composed of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a protein of interest. They function by exploiting the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Utilizes the intracellular ubiquitin-proteasome system