Learn More
Description
- 1,2,4-butanetriol was used as starting reagent during the synthesis of a series of quaternary ammonium lipids
- It was also used in the synthesis of (-)-α-amino-8-hydroxy butyric acid (gabob), antiepileptic and hypotensive drug
Specifications
Specifications
| CAS | 3068-00-6 |
| Density | 1.184 g/mL at 20°C, 1.19 g/mL (at 25°C (literature)) |
| Boiling Point | 190°C to 191°C (18 mmHg) |
| Molecular Formula | C4H10O3 |
| Refractive Index | n20/D 1.473 (literature); n20/D 1.473 |
| Linear Formula | HOCH2CH2CH(OH)CH2OH |
| MDL Number | MFCD00002929 |
| Quantity | 100 mL |
| Molecular Weight (g/mol) | 106.12 |
| Percent Purity | ≥90% (GC) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.