Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+/-)-1,2,4-Butanetriol

Catalog No. 1904025ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-040-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-040-25ML Supplier Sigma Aldrich Supplier No. 1904025ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2,4-butanetriol was used as starting reagent during the synthesis of a series of quaternary ammonium lipids
  • It was also used in the synthesis of (-)-α-amino-8-hydroxy butyric acid (gabob), antiepileptic and hypotensive drug

Specifications

CAS 3068-00-6
Density 1.184 g/mL at 20°C, 1.19 g/mL (at 25°C (literature))
Boiling Point 190°C to 191°C (18 mmHg)
Molecular Formula C4H10O3
Refractive Index n20/D 1.473 (literature); n20/D 1.473
Linear Formula HOCH2CH2CH(OH)CH2OH
MDL Number MFCD00002929
Quantity 25 mL
Molecular Weight (g/mol) 106.12
Percent Purity ≥90% (GC)
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.