Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Butanetriol, 96%, Thermo Scientific Chemicals

Catalog No. AAL0513814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0513814 25 g
AAL0513806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0513814 Supplier Thermo Scientific Chemicals Supplier No. L0513814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3068-00-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.121
MDL Number MFCD00002929
InChI Key ARXKVVRQIIOZGF-UHFFFAOYSA-N
Synonym 1,2,4-butanetriol, 2-deoxyerythritol, 1,3,4-butanetriol, triol 124, 1,2,4-trihydroxybutane, 1,2,4-butantriol, 1,2,4-butantriol german, +/--1,2,4-butanetriol, 2-deoxyerthritol, dialkylenethiourea
PubChem CID 18302
IUPAC Name butane-1,2,4-triol
SMILES C(CO)C(CO)O

Specifications

Melting Point -20°C
Density 1.182
Boiling Point 190°C to 191°C (18 mmHg)
Flash Point 188°C (370°F)
Refractive Index 1.475
Quantity 25 g
Beilstein 1733456
Sensitivity Hygroscopic
Formula Weight 106.12
Percent Purity 96%
Chemical Name or Material 1,2,4-Butanetriol
Show More Show Less
EINECSNumber 221-323-5
RTECSNumber EK7176000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.