Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. D362925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D362925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D362925G Supplier TCI America Supplier No. D362925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24155-42-8
Molecular Formula C11H10Cl2N2O
Molecular Weight (g/mol) 257.114
MDL Number MFCD00044708
InChI Key UKVLTPAGJIYSGN-UHFFFAOYSA-N
Synonym alpha-2,4-dichlorophenyl-1h-imidazole-1-ethanol, 1-2,4-dichlorophenyl-2-1h-imidazol-1-yl ethanol, 1-2,4-dichlorophenyl-2-1h-imidazol-1-yl ethan-1-ol, 1-2,4-dichlorophenyl-2-1-imidazolyl ethanol, chembl1327, 1-2,4-dichlorophenyl-2-imidazol-1-ylethanol, 1h-imidazole-1-ethanol, alpha-2,4-dichlorophenyl, 1-2,4-dichloro-phenyl-2-imidazol-1-yl-ethanol, benzyl alcohol, 2,4-dichloro-alpha-1-imidazolylmethyl, n-beta-2,4-dichlorophenyl-beta-hydroxyethyl imidazole
PubChem CID 32238
IUPAC Name 1-(2,4-dichlorophenyl)-2-imidazol-1-ylethanol
SMILES C1=CC(=C(C=C1Cl)Cl)C(CN2C=CN=C2)O

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 25 g
Formula Weight 257.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol
RTECSNumber NI5485000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.