Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Difluorophenyl)piperazine, 99%, Thermo Scientific™

Catalog No. AAL1939303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1939303 1 g
AAL1939306 5 g
AAL1939314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1939303 Supplier Thermo Scientific Chemicals Supplier No. L1939303
  • $46.50
    $41.65 / Each of 1

    Save $4.85

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115761-79-0
Molecular Formula C10H12F2N2
Molecular Weight (g/mol) 198.217
MDL Number MFCD00082588
InChI Key CMCSPBOWEYUGHB-UHFFFAOYSA-N
PubChem CID 2734637
IUPAC Name 1-(2,4-difluorophenyl)piperazine
SMILES C1CN(CCN1)C2=C(C=C(C=C2)F)F

Specifications

Melting Point 74°C to 76°C
Boiling Point 108°C to 111°C (0.2 mmHg)
Quantity 1 g
UN Number UN2923
Beilstein 8620369
Sensitivity Air Sensitive
Synonym 1-2,4-difluorophenyl piperazine, 1-2,4-difluorophenyl-piperazine, piperazine, 1-2,4-difluorophenyl, 2,4-difluorophenyl piperazine, pubchem9352, acmc-1ca8h, 2,4-difluorophenylpiperazine, 4-2,4-difluorophenyl piperazine, 1-2,4-difluoro-phenyl-piperazine, piperazine,1-2,4-difluorophenyl
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 1-(2,4-Difluorophenyl)piperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.