Learn More
Sigma Aldrich 1-(2,4-Dihydroxyphenyl)-3-(4-ethylphenoxy)-1-propanone
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Cyp2c19 metabolite of mephenytoin.pharmacological activity investigating:
- The drug effects on metabolization
- In vitro cytochrome p450 activity in microsomesanalyte for electron-capture gas chromatography procedures
Specifications
Specifications
| Molecular Formula | C17H18O4 |
| Linear Formula | C17H18O4 |
| MDL Number | MFCD09057541 |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 286.32 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.