Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triaminobenzene dihydrochloride, 96%, Thermo Scientific™

Catalog No. AAA1775806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1775806 5 g
AAA1775814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1775806 Supplier Thermo Scientific Chemicals Supplier No. A1775806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-47-4
Molecular Formula C6H11Cl2N3
Molecular Weight (g/mol) 196.08
MDL Number MFCD00016619
InChI Key RKMFFOLUBJFMBQ-UHFFFAOYSA-N
Synonym 1,2,4-benzenetriamine dihydrochloride, 1,2,4-triaminobenzene dihydrochloride, benzene-1,2,4-triamine dihydrochloride, 1,2,4-benzenetriamine, dihydrochloride, benzene-1,2,4-triyltriamine dihydrochloride, 1,2,4-benzenetriamine, hydrochloride 1:2, benzene-1,2,4-triamine, chloride, chloride, benzol-1,2,4-triamindihydrochlorid, 1,2,4-benzenetriaminedihydrochloride, acmc-1bb7p
PubChem CID 94179
IUPAC Name benzene-1,2,4-triamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C(N)=C1

Specifications

Melting Point ∼290°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 196.08
Percent Purity 96%
Chemical Name or Material 1,2,4-Triaminobenzene dihydrochloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 210-428-1
RTECSNumber DC1953000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.