Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triaminobenzene Dihydrochloride 95.0+%, TCI America™
SDP

Catalog No. T03341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T03341G Supplier TCI America Supplier No. T03341G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 615-47-4
Molecular Formula C6H11Cl2N3
Molecular Weight (g/mol) 196.08
MDL Number MFCD00016619
InChI Key RKMFFOLUBJFMBQ-UHFFFAOYSA-N
Synonym 1,2,4-benzenetriamine dihydrochloride, 1,2,4-triaminobenzene dihydrochloride, benzene-1,2,4-triamine dihydrochloride, 1,2,4-benzenetriamine, dihydrochloride, benzene-1,2,4-triyltriamine dihydrochloride, 1,2,4-benzenetriamine, hydrochloride 1:2, benzene-1,2,4-triamine, chloride, chloride, benzol-1,2,4-triamindihydrochlorid, 1,2,4-benzenetriaminedihydrochloride, acmc-1bb7p
PubChem CID 94179
IUPAC Name dihydrogen benzene-1,2,4-triamine dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C(N)=C1

Specifications

Quantity 1 g
Formula Weight 196.08
Percent Purity ≥95.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triaminobenzene Dihydrochloride
RTECSNumber DC1953000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.