Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,4-Triazin-3-amine | 1120-99-6 | 96.09 | 1 ML
SDP

Catalog No. 5000446067
Encompass_Preferred

3-Amino-1,2,4-triazine is a chemical compound known for its inhibitory effects on NO synthase and nitrite secretion. It is provided as a solution and is intended for research use.

  • Inhibits NO synthase activity in cytosolic extracts of activated cells.
  • Inhibits both nitrite secretion and parasite killing by activated macrophages.
  • Molecular formula: C3H4N4.
  • Purity: 98.0%.
  • Molecular weight: 96.09.
  • Appearance: Brown to black solid.
  • Store at 4°C, protected from light.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-004-46067 Supplier Medchemexpress LLC Supplier No. HYW00484310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Amino-1,2,4-triazine is a chemical compound known for its inhibitory effects on NO synthase and nitrite secretion. It is provided as a solution and is intended for research use.

  • Inhibits NO synthase activity in cytosolic extracts of activated cells.
  • Inhibits both nitrite secretion and parasite killing by activated macrophages.
  • Molecular formula: C3H4N4.
  • Purity: 98.0%.
  • Molecular weight: 96.09.
  • Appearance: Brown to black solid.
  • Store at 4°C, protected from light.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.